missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum 13 wt% Rhodium wire, 0.076mm (0.003 in.) dia., ISA Type R Standard Grade Thermocouple

Brand:  Alfa Aesar™ 042902.G6

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 10m

 View more versions of this product

Product Code. 11300158

  • £509.30 / 10m


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Platinum 13 wt% Rhodium wire
0.076mm (0.003 in.) dia., ISA Type R Standard Grade Thermocouple
MFCD00198186

Specifications

Pt:Rh; 87:13 wt%
≈0.045g/50cm
0.076mm (0.003 in.) dia., ISA Type R Standard Grade Thermocouple
MFCD00198186
Platinum 13 wt% Rhodium wire
10m
Wire
Odorless
Videos
SDS