Learn More
Platinum 10 wt% Rhodium wire, 0.076mm (0.003in) dia, ISA Type S Standard Grade Thermocouple, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 042891.BW
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum 10 wt% Rhodium wire | |
Pt:Rh; 90:10 wt% | |
50 cm | |
≈0.046g/50cm |
0.076mm (0.003 in.) dia., ISA Type S Standard Grade Thermocouple | |
MFCD00198186 | |
Odorless | |
Wire |
RUO – Research Use Only