missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum 10 wt% Rhodium wire, 0.076mm (0.003 in.) dia., ISA Type S Standard Grade Thermocouple

Brand:  Alfa Aesar™ 042891.G6

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 10m

 View more versions of this product

Product Code. 11320138

  • POA / 10m

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

Platinum 10 wt% Rhodium wire
0.076mm (0.003 in.) dia., ISA Type S Standard Grade Thermocouple
MFCD00198186

Specifications

Pt:Rh; 90:10 wt%
≈0.046g/50cm
0.076mm (0.003 in.) dia., ISA Type S Standard Grade Thermocouple
MFCD00198186
Platinum 10 wt% Rhodium wire
10m
Wire
Odorless
Videos
SDS