missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 1% on activated carbon powder, standard, unreduced, nominally 50% water wet, Thermo Scientific™

Catalog Number p-7050441
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7440-6-6
MFCD00011179
This item is not returnable. View return policy
7440-6-6
MFCD00011179

Schiff's base and nitrohydrogenation, hydrosilation

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Solubility Information Soluble in aqua regia.. Insoluble in water and acid.
Chemical Name or Material Platinum, 1% on activated carbon powder, Standard, unreduced, nominally 50% water wet
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'