missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum, 1% on activated carbon powder, standard, reduced, nominally 50% water wet

Brand:  Alfa Aesar™ 038312.22

Code : NEW

Additional Details : Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11304437

  • £435.10 / 100g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Platinum
1% on activated carbon powder, Standard, reduced, nominally 50% water wet
MFCD00011179

Specifications

Platinum
7440-6-6
100g
1% on activated carbon powder, Standard, reduced, nominally 50% water wet
MFCD00011179
900 to 1100m2/g
Videos
SDS