missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum, 1% on activated carbon paste, 1R18MA

Brand:  Alfa Aesar™ 045519.14

Code : NEW

Additional Details : CAS Number : 7440-06-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15402337

  • £152.00 / 25g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

Platinum
7440-06-4
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Paste
1% on activated carbon paste, 1R18MA
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
25g
Videos
SDS Documents
Certificates