missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum, 0.5% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced

Brand:  Alfa Aesar™ 089106.14

Code : NEW

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11309389

  • £56.70 / 25g


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Description

Description

  • Selective hydrogenations; gas purifications: e
g
  • Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
  • Specifications

    Chemical Identifiers

    Platinum
    0.5% on alumina pellets, 2.7 to 3.3mm (0.11 to 0.13 in.), Reduced
    MFCD00011179

    Specifications

    Platinum
    0.5% on alumina pellets, 2.7 to 3.3mm (0.11 to 0.13 in.), Reduced
    MFCD00011179
    25g
    Videos
    SDS