missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 0.3% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced, Thermo Scientific Chemicals

Catalog Number p-7050953
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7440-6-6
MFCD00011179
This item is not returnable. View return policy
7440-6-6
MFCD00011179

Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
TRUSTED_SUSTAINABILITY
Quantity 25 g
Chemical Name or Material Platinum, 0.3% on 2.7 to 3.3mm (0.11 to 0.13 in.) alumina pellets, Reduced

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'