missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum, 0.1% on 2-4mm alumina spheres

VOC removal from vent streams by combination with O2

Brand:  Alfa Aesar™ 042205.22

Code : NEW

Additional Details : Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11349857

  • £517.70 / 100g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Platinum
0.1% on 2 to 4mm alumina spheres
MFCD00011179

Specifications

Platinum
Spheres
100g
0.1% on 2 to 4mm alumina spheres
MFCD00011179
Videos
SDS