Learn More
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators
Accommodates tubes of different diameters. Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators keeps test tubes secure during rotation
Brand: Grant Instruments™ PRS-8-22
Code : 04
Additional Details : Weight : 0.50000kg
Description
- For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
- Hold as few as 8 test tubes or as many as 48.
- Accommodates a variety of capacities
Specifications
8 tubes up to 30mm plus 22 tubes up to 15mm, tube volumes up to 50mL/1.5-15mL | |
8 tubes up to 30 mm plus 22 tubes up to 15 mm, tube volumes up to 50 mL/1.5-15 mL |