missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platelet-poor Plasma Derived Serum, Bovine

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J64483.AB

Code : NEW

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15406419

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Format: Liquid
Specifications

Specifications

Liquid
Bovine
-30° to -10°C
Sterile-filtered
10mL
Platelet-poor Plasma Derived Serum, Bovine
Videos
SDS

For Research Use Only