missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ Rigid 1mm Plastic PVC Safety Signs

Follow best practices regarding potential hazards by posting Allsigns International™ Rigid 1mm Plastic PVC Safety Signs in areas that may pose a danger to public safety.

Brand:  Allsigns International™ WA124R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12305658

  • POA / Each


Estimated Shipment
14-06-2017
Add to basket

Specifications

Description

  • Black triangle symbol and text, yellow background
  • Self-adhesive
  • 1mm PVC rigid plastic
Description & Specifications

Specifications

200mm x 300mm
Caution Hot Surface
Chemicals
Rigid PVC Plastic
Rectangle