Allsigns International™ Rigid 1mm Plastic PVC Safety Signs

Follow best practices regarding potential hazards by posting Allsigns International™ Rigid 1mm Plastic PVC Safety Signs in areas that may pose a danger to public safety.

Overview
Brand: Allsigns International™

Manufacturer Part Number: WA217R

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12365698

Quantity Price
1 £ 21.72 / Each
Estimated Shipment
08-06-2017
Add to basket


Description and Specification

Specification

Dimensions (L x W) 200mm x 300mm
Material Rigid PVC Plastic
Message Hazardous Waste
Shape Rectangle
Resistant To Chemicals

  • Black triangle symbol and text, yellow background
  • Self-adhesive
  • 1mm PVC rigid plastic

Documentation