Allsigns International™ Plastic Safety Sign Label


Overview
Brand: Allsigns International™

Manufacturer Part Number: XJR-451-P

Safety sign CO3/PVC multi purpose sign dangerhazardous substances 400mm x 300mm 1mm rigid

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12385528

Quantity Price
1 £ 24.41 / Each
Estimated Shipment
30-03-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Used to Instruct, Advise and Forewarn Staff and Visitors of Potential Dangers Arising In and Around the Workplace
Width (Metric) 300mm
Quantity 1Each
Adhesive Type Self
Type Safety Sign Label
Resistant To Chemicals
Material Rigid Plastic
Message Danger Hazardous Substances Please Observe the Correct Precautions
Dimensions (L x W) 400 x 300mm
Thickness 1mm
Shape Rectangle
Length (Metric) 400mm
Documentation

Technical documents