Allsigns International™ Plastic Safety Sign Label


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA157/PVC.

Safety sign WA157/PVC hazard warning danger acid200mm x 300mm 1mm rigid plastic Allsigns, 1/Cs,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12305698

Quantity Price
1 £ 21.97 / Each
Estimated Shipment
06-03-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Used to Instruct, Advise and Forewarn Staff and Visitors of Potential Dangers Arising In and Around the Workplace
Width (Metric) 300mm
Adhesive Type Self
Quantity 1Each
Type Safety Sign Label
Resistant To Chemicals
Color Black Markings with Yellow on White Background
Material Rigid Plastic
Message Danger Acid
Dimensions (L x W) 200 x 300mm
Thickness 1mm
Shape Rectangle
Length (Metric) 200mm

To instruct, advise and forewarn staff and visitors of potential dangers arising in and around the workplace. Color - black symbol and text, yellow backgroundSymbol - triangleS/A - Self-Adhesive

Documentation