missing translation for 'onlineSavingsMsg'
Learn More

Abritech™ Plastic Resistance Coil

Brand:  Abritech™ XJH-840-190T

Code : 02

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13166460

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Resistance coil
With two 4mm socket
Plastic
5ohms
1/Ea.
Videos
Documentation

Technical Documents