Learn More
Fisherbrand™ Plastic Cover with Orifice
Allows glass beakers with a diameter between 90 and 97mm to be inserted into the ultrasonic bath for direct treatment. Fisherbrand™ Plastic Covers with Orifice can be combined with rubber rings to securely and safely hold the beaker during the ultrasonic treatment process.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.18900kg
Description
- Manufactured from durable, blue plastic
- For use with S 50 R Ultrasonic Cleaning Units
Specifications
Lid | |
Beaker | |
Plastic |