missing translation for 'onlineSavingsMsg'
Learn More

Manutan Plastic divider, 21 compartment, height 100mm to fit 650mm x 650mm

Brand:  Manutan A348666

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12808138

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Fully customisable drawer divider inserts. Choose from adjustable galvanised sheet steel or fixed moulded dark grey polystyrol.
  • 21 compartments
  • Drawer O/A height: 100mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers