missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Plasmid DNA purification, NucleoBond™ , Resuspension Buffer RES
Brand: Macherey-Nagel™ 740363.1000
70.95 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.
Code : LE
Additional Details : Weight : 1.00000kg
Specifications
1000 mL | |
For Plasmid DNA Purification | |
Resuspension Buffer |