missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Kallikrein/KLKB1 Rat anti-Mouse, Clone: 337801, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Plasma Kallikrein/KLKB1 |
---|---|
Clone | 337801 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Kallikrein/KLKB1 Gly20-Ala638 Accession # NP_032481 |
Host Species | Rat |
Primary or Secondary | Primary |
Description
Plasma Kallikrein/KLKB1 Monoclonal specifically detects Plasma Kallikrein/KLKB1 in Mouse samples. It is validated for Western Blot.Specifications
Plasma Kallikrein/KLKB1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Kallikrein/KLKB1 Gly20-Ala638 Accession # NP_032481 | |
Primary | |
Mouse | |
Unconjugated | |
3818 | |
Purified | |
Western Blot | |
RUO |
337801 | |
Rat | |
Monoclonal | |
IgG1 | |
EC 3.4.21, EC 3.4.21.34, Fletcher factor, kallikrein B, plasma (Fletcher factor) 1, kininogenin, KLK3plasma kallikrein, KLKB1, plasma kallikrein heavy chain, plasma kallikrein light chain, Plasma prekallikrein, PPK | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only