missing translation for 'onlineSavingsMsg'
Learn More

Motic™ CCIS™ EC-H Plan Phase Objective, Negative Phase

CCIS™ EC-H Plan Phase Objective, Negative Phase is compatible with all BA series microscopes

Brand:  Motic™ 1101001703421

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13425509

  • £445.00 / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

  • For use with BA Series Microscopes
  • EC-H Plan Phase
  • Negative Phase
  • Magnifications/Numerical Apertures available:
    • 10X/0.25 (WD=17.4mm)
    • 20X/0.45 (WD=0.9mm)
    • 40X/0.65/S (WD=0.5mm)
    • 100X/1.25/S-Oil (WD=0.15mm)
Specifications

Specifications

CCIS, Plan Achromatic Phase Objective, Negative Phase
0.15 WD
100X
1 Ea.
EC-H Plan Phase
BA Microscope Series
1.25
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers