missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Plan-Achromat Objective

Designed for use with Carl Zeis Primo Star Microscopes

Brand:  Zeiss™ 415500-1620-000

Code : 03

Additional Details : Weight : 0.13000kg

Product Code. 15150143

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Objective Plan-Achromat 100x/0.8 dry (WD: 0.94mm)
  • Compatible Microscopes:

    • Primo Star
    • Primo Star iLED
Specifications

Specifications

Objective Planachromatic
0.94 mm
100X
100x/0.8 dry for Primo
Primo Microscopes
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers