missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550642
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total PLA2G4B protein.
PLA2G4B Polyclonal antibody specifically detects PLA2G4B in Human, Mouse samples. It is validated for Western Blot.Specifications
PLA2G4B | |
Polyclonal | |
Unconjugated | |
PLA2G4B | |
A030011C02Rik; cPLA2-beta; cytosolic phospholipase A2 beta; Gm1013; HsT16992; JMJD7-PLA2G4B; JMJD7-PLA2G4B protein; JMJD7-PLA2G4B readthrough; jumonji domain containing 7-phospholipase A2, group IVB (cytosolic) read-through; phospholipase A2 group IVB; phospholipase A2, group IVB (cytosolic); Pla2g4b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100137049, 211429 | |
-20°C | |
Liquid |
Western Blot | |
2.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P0C869, P0C871 | |
PLA2G4B | |
Synthetic peptide corresponding to a region derived from internal residues of human phospholipase A2, group IVB (cytosolic) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |