missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKN2 (PRK2) Recombinant Human Protein
PKN2 (PRK2) Protein
Brand: Invitrogen PV3879
Code : 50
Additional Details : Weight : 0.05250kg
Description
PKN2 (PRK2), a close relative of the PKCs, is a regulator of mitosis and exit from cytokinesis.Specifications
NP_006247 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
141.5kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
PKN2 (PRK2) | |
PKN2 (PRK2) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 533μg/mL MAPKAPK2 peptide substrate (KKLNRTLSVA) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
PKN2 (PRK2) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.29mg/mL | |
114294, 5586 | |
PKN2 (PRK2) Recombinant Human Protein | |
–80°C | |
1 Each | |
PKN2 (PRK2) Recombinant Human Protein | |
PKN | |
18nmoles of phosphate transferred to MAPKAPK2 peptide substrate (KKLNRTLSVA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 16.7μg/mL | |
GST Tag | |
Kinase and inhibitors | |
80% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.