missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIWIL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA568430
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse samples.
PIWIL2 Polyclonal antibody specifically detects PIWIL2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
PIWIL2 | |
Polyclonal | |
Unconjugated | |
Piwil2 | |
80kDa PIWIL2 short isoform; cancer/testis antigen 80; CT80; HILI; mili; Miwi like; Miwi likw; piwi like homolog 2; piwi like RNA-mediated gene silencing 2; PIWIL1L; Piwil2; PIWI-L2; piwil2-like protein; piwi-like 2; piwi-like homolog 2; piwi-like protein 2; piwi-like RNA-mediated gene silencing 2; testicular tissue protein Li 141 | |
Rabbit | |
affinity chromatography | |
RUO | |
55124, 57746 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8CDG1, Q8TC59 | |
Piwil2 | |
Synthesized peptide derived from human PIWIL2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |