missing translation for 'onlineSavingsMsg'
Learn More

Pivaloylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-3779498
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
59997-51-2
C7H11NO
125.17
MFCD00010208
MXZMACXOMZKYHJ-UHFFFAOYSA-N
pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
108871
4,4-dimethyl-3-oxopentanenitrile
CC(C)(C)C(=O)CC#N
This item is not returnable. View return policy
59997-51-2
C7H11NO
125.17
MFCD00010208
MXZMACXOMZKYHJ-UHFFFAOYSA-N
pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
108871
4,4-dimethyl-3-oxopentanenitrile
CC(C)(C)C(=O)CC#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 66°C to 72°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC(O)CH2CN
Quantity 25 g
Formula Weight 125.17
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Pivaloylacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • Pivaloylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'