missing translation for 'onlineSavingsMsg'
Learn More

Pivaloylacetonitrile, 97%, ACROS Organics™

Brand:  Acros Organics 418025000

Code : 29

Additional Details : CAS Number : 59997-51-2 Weight : 0.55000kg Transport : UN number : 3439 Chem class : 1 Pack group : III


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10557562

  • £422.80 / 500g


Estimated Shipment
23-05-2018
Add to basket


Specifications

Chemical Identifiers

Pivaloylacetonitrile
97%
(CH3)3CC(O)CH2CN
MFCD00010208
≥96.0%
C7H11NO
125.17

Specifications

Pivaloylacetonitrile
Crystals or Crystalline Powder
97%
C7H11NO
125.17
Glass bottle
White to cream
500g
59997-51-2
≥96.0%
Authentic
(CH3)3CC(O)CH2CN
MFCD00010208
1% max. (K.F.)
66° to 72°C
Videos
SDS