missing translation for 'onlineSavingsMsg'
Learn More

Piribedil dihydrochloride, Tocris Bioscience™

Dopamine agonist

Brand:  Tocris (Bio-Techne) 103110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13458343

  • £128.25 / 10mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

A direct dopamine agonist, in clinical use for treatment of dopaminergic system dysfunction. Recent work suggests that it is selective for the D3 subtype, for which it has 20 times higher affinity than for D2, and possesses no significant affinity for D1 receptors.
Specifications

Chemical Identifiers

1451048-94-4
371.262
piribedil dihydrochloride, 2-4-1,3-benzodioxol-5-ylmethyl-1-piperazinyl pyrimidine dihydrochloride
2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]pyrimidine;dihydrochloride
C16H20Cl2N4O2
LTXWEVPOJXPWAD-UHFFFAOYSA-N
56972173
C1CN(CCN1CC2=CC3=C(C=C2)OCO3)C4=NC=CC=N4.Cl.Cl

Specifications

Piribedil dihydrochloride
1451048-94-4
piribedil dihydrochloride, 2-4-1,3-benzodioxol-5-ylmethyl-1-piperazinyl pyrimidine dihydrochloride
C1CN(CCN1CC2=CC3=C(C=C2)OCO3)C4=NC=CC=N4.Cl.Cl
371.262
371.27
10mg
C16H20Cl2N4O2
LTXWEVPOJXPWAD-UHFFFAOYSA-N
2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]pyrimidine;dihydrochloride
56972173
>99%
Videos