Learn More
BRAND™ Natural Rubber Pipet Filler
Manufactured from 100% natural rubber. BRAND™ Natural Rubber Pipet Filler eliminates mouth pipetting and handling of hazardous material. Available in universal, standard, or flip model, with 2 or 3-valve options.
Brand: BRAND™ 25400
12.33 GBP valid until 2024-03-30
Use promo code "21680" to get your promotional price.
Code : A0
Additional Details : Weight : 0.04500kg
Description
- Manufactured from 100% natural rubber.
- Eliminates mouth pipetting and handling of hazardous material.
- Available in universal, standard, or flip model, with 2 or 3-valve options
- 2-valve flip model enables one-handed operation.
Specifications
Valve ↑: Filling, Valve ↓: Delivery, Releases air through an automatic valve | |
Flip | |
Filling pipets up to 100mL | |
1 Each |
Natural Rubber | |
100 mL | |
2 valves |