Gilson™ Pipetman™ G 2nd Level Service Kit
Overview
Brand: Gilson™
Manufacturer Part Number: F1619742
service kit 2nd level P200G
UNSPSC: 73152100
Code: 50
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 12988290
Quantity Price
1 £ 16.310000000000002 / Each
Estimated Shipment
15-03-2017
Add to basket
Description and Specification
Specification
For Use With (Equipment) For Metal and Plastic PIPETMAN P200G
Packaging Quantity 1Kit
Product Type PIPETMAN G 2nd Level Service Kit
Includes 1 Push-button, 1 connecting nut, 1 Stainless Steel ejector, 1 Plastic ejector, 2 ejector adapters