Gilson™ Pipetman™ G 1st Level Service Kit
Overview
Brand: Gilson™
Manufacturer Part Number: FA07001
service kit, 1st level P10G/P10L
UNSPSC: 73152100
Code: 50
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 12998300
Quantity Price
1 £ 51.71 / Each
Estimated Shipment
09-02-2017
Add to basket
Description and Specification
Specification
For Use With (Equipment) For Pipetman P10G
Packaging Quantity 1Kit
Product Type Pipetman G 1st Level Service Kit
Includes 3 Piston seals or seal guides, 3 o-rings, 1 tip-holder