Learn More
Fisherbrand™ Top-Line Pipette Tips
Eliminate sample contamination and prevent sample loss. . Fisherbrand™ Top-Line Pipette Tips provide precision pipetting with conical tip and secure fit.
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 0.96000kg
 View more versions of this product
Product Code. 10396142
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Optimized cone geometry for a secure fit
  • Graduation marks on tip at standard volumes
  • Conical tip opening prevents drop formation at the end of the tip
  • Tips made of autoclavable, non-wettable, virgin polypropylene
  • Sterile packs available (beta irradiation)
Specifications
Specifications
5 to 300 μL
Clear
55mm
Autoclave
Universal
Yes
Virgin Polypropylene
Non-sterile
Autoclavable
ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600
Rack
Universal Fit
Top-Line
Standard volumes