missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, Ultrapure Bioreagent, J.T. Baker™
196.65 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Specifications
CAS | 5625-37-6 |
Quantity | 100 g |
Molecular Formula | C8H18N2O6S2 |
Packaging | Poly Bottle |
Synonym | pipes, 1,4-piperazinediethanesulfonic acid, piperazine-n,n'-bis 2-ethanesulfonic acid, 2,2'-piperazine-1,4-diyl diethanesulfonic acid, 1,4-piperazinebis ethanesulfonic acid, unii-g502h79v6l, 2-4-2-sulfoethyl piperazin-1-yl ethanesulfonic acid, piperazine-1,4-bis 2-ethanesulfonic acid, 1,4-piperazine-diethanesulfonic acid, 2,2'-piperazine-1,4-diyl bis ethanesulphonic acid |
InChI Key | IHPYMWDTONKSCO-UHFFFAOYSA-N |
SMILES | C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
IUPAC Name | 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
Molecular Weight (g/mol) | 302.36 |
PubChem CID | 79723 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'