missing translation for 'onlineSavingsMsg'
Learn More

Piperonylonitrile, 97%, Thermo Scientific™

Catalog Number p-185513
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4421-09-4
147.13
MFCD00005820
PKRWWZCDLJSJIF-UHFFFAOYSA-N
piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358
78136
1,3-benzodioxole-5-carbonitrile
C1OC2=C(O1)C=C(C=C2)C#N
This item is not returnable. View return policy
4421-09-4
147.13
MFCD00005820
PKRWWZCDLJSJIF-UHFFFAOYSA-N
piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358
78136
1,3-benzodioxole-5-carbonitrile
C1OC2=C(O1)C=C(C=C2)C#N
Melting Point 90.0°C to 95.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H5NO2
Quantity 5 g
Beilstein 19, 270
Formula Weight 147.13
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Piperonylonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Piperonylonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'