missing translation for 'onlineSavingsMsg'
Learn More

Piperine, 98%, Thermo Scientific Chemicals

Catalog Number p-3213059
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
94-62-2
C17H19NO3
285.34
MFCD00005839
MXXWOMGUGJBKIW-YPCIICBESA-N
piperine, 1-piperoylpiperidine, piperin, bioperine, piperoylpiperidine, e,e-1-piperoylpiperidine, n-e,e-piperoyl piperidine, piperidine, 1-piperoyl-, e,e, unii-u71xl721qk, fema no. 2909
638024
CHEBI:28821
(2E,4E)-5-(1,3-benzodioxol-5-yl)-1-piperidin-1-ylpenta-2,4-dien-1-one
O=C(\C=C\C=C\C1=CC=C2OCOC2=C1)N1CCCCC1
94-62-2
C17H19NO3
285.34
MFCD00005839
MXXWOMGUGJBKIW-YPCIICBESA-N
piperine, 1-piperoylpiperidine, piperin, bioperine, piperoylpiperidine, e,e-1-piperoylpiperidine, n-e,e-piperoyl piperidine, piperidine, 1-piperoyl-, e,e, unii-u71xl721qk, fema no. 2909
638024
CHEBI:28821
(2E,4E)-5-(1,3-benzodioxol-5-yl)-1-piperidin-1-ylpenta-2,4-dien-1-one
O=C(\C=C\C=C\C1=CC=C2OCOC2=C1)N1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 128.0°C to 132.0°C
Color Tan to White or Tan-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 7584
Solubility Information Solubility in water: 40 mg/l (18°C). Other solubilities: 1g/15mL alcohol - 1g/1.7mL chloroform, 1g/36mL ether, soluble in benzene and acetic acid, practically insoluble in petroleum ether
Formula Weight 285.34
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Piperine
Show More Show Less
compliance-icons
Product Identifier
  • Piperine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'