missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperidino(4-piperidinyl)methanone, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | piperidin-1-yl(piperidin-4-yl)methanone |
Formula Weight | 196.29 |
Percent Purity | 97% |
Chemical Name or Material | Piperidino(4-piperidinyl)methanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'