missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Piperazine hydrogen phosphate monohydrate, 98+%

Brand:  Alfa Aesar™ A11487.22

Code : NEW

Additional Details : CAS Number : 18534-18-4 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11470383

  • £24.30 / 100g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

Piperazine hydrogen phosphate monohydrate
C4H10N2·H3PO4·H2O
MFCD00156159
18534-18-4
202.14 (184.12 Anhydrous)

Specifications

Piperazine hydrogen phosphate monohydrate
18534-18-4
C4H10N2·H3PO4·H2O
MFCD00156159
100g
≥98%
202.14 (184.12 Anhydrous)
Videos
SDS