missing translation for 'onlineSavingsMsg'
Learn More

Piperazine hexahydrate, 98%, ACROS Organics™

Brand:  Acros Organics 131300010

Code : 29

Additional Details : CAS Number : 142-63-2 Weight : 1.05000kg Transport : UN number : 2579 Chem class : 8 Pack group : III


Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10795611

  • £57.80 / 1kg


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

Piperazine hexahydrate
142-63-2
98%
194.23
98%
98%
C4H10N2·6H2O
MFCD00149389

Specifications

Piperazine hexahydrate
145.0° to 156.0°C
41.0° to 45.0°C
98%
Authentic
Adhering Crystals
98%
194.23
87°C
White or transparent
1kg
142-63-2
Plastic bottle
98%
C4H10N2·6H2O
MFCD00149389
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Piperazine hexahydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS