missing translation for 'onlineSavingsMsg'
Learn More

Pioglitazone hydrochloride, Tocris Bioscience™

Selective PPARγ agonist; antidiabetic agent

Brand:  Tocris (Bio-Techne) 412450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13464194

  • £249.49 / 50mg


Estimated Shipment
03-01-2019
Add to basket

Description

Description

Selective PPARγ agonist (EC50 = 0.69μM). Thiazolidinedione (TZD) derivative and antidiabetic agent; improves insulin sensitivity.
Specifications

Chemical Identifiers

112529-15-4
392.898
pioglitazone hydrochloride, pioglitazone hcl, 5-4-2-5-ethylpyridin-2-yl ethoxy benzyl thiazolidine-2,4-dione hydrochloride, pioglitazone hydrochloride, pioditazone hydrochloride, 5-4-2-5-ethyl-2-pyridinyl ethoxy phenyl methyl-2,4-thiazolidinedione hydrochloride, +--5-p-2-5-ethyl-2-pyridyl ethoxy benzyl-2,4-thiazolidinedione monohydrochloride, dsstox_cid_24203, dsstox_rid_80116, dsstox_gsid_44203
5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride
C19H21ClN2O3S
GHUUBYQTCDQWRA-UHFFFAOYSA-N
60560
CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl

Specifications

Pioglitazone hydrochloride
112529-15-4
pioglitazone hydrochloride, pioglitazone hcl, 5-4-2-5-ethylpyridin-2-yl ethoxy benzyl thiazolidine-2,4-dione hydrochloride, pioglitazone hydrochloride, pioditazone hydrochloride, 5-4-2-5-ethyl-2-pyridinyl ethoxy phenyl methyl-2,4-thiazolidinedione hydrochloride, +--5-p-2-5-ethyl-2-pyridyl ethoxy benzyl-2,4-thiazolidinedione monohydrochloride, dsstox_cid_24203, dsstox_rid_80116, dsstox_gsid_44203
CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl
392.898
392.9
50mg
C19H21ClN2O3S
GHUUBYQTCDQWRA-UHFFFAOYSA-N
5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride
60560
>99%
Videos