missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIN Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | PIN/DLC8 |
---|---|
Immunogen | E. coli-derived recombinant human PIN Trp2-Gly89 Accession # P63167 |
Gene ID (Entrez) | 8655 |
Gene Accession No. | P63167 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
PIN/DLC8 Polyclonal specifically detects PIN/DLC8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
PIN/DLC8 | |
8655 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human PIN in direct ELISAs and Western blots. | |
Unconjugated | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
E. coli-derived recombinant human PIN Trp2-Gly89 Accession # P63167 | |
P63167 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only