missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILR-beta Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | PILR-beta |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse PILR-beta Ala28-Gly193 Accession # Q2YFS2 |
Gene ID (Entrez) | 29990 |
Gene Accession No. | Q2YFS2 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
PILR-beta Polyclonal specifically detects PILR-beta in Mouse samples. It is validated for Western Blot.Specifications
PILR-beta | |
29990 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse PILR-beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse PILR-alpha is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant mouse PILR-beta Ala28-Gly193 Accession # Q2YFS2 | |
Q2YFS2 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only