missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PIK3C3 (hVPS34), GST Tag Recombinant Protein
Human PIK3C3 (hVPS34), GST Tag Recombinant Protein
£564.00 - £12060.00
Specifications
Accession Number | NP_002638.2 |
---|---|
Gene Alias | PIK3C3 (hVPS34) |
KinaseFamily | PI3 Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448703
|
Invitrogen™
PR8987B |
100μg |
£2555.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498773
|
Invitrogen™
PV5127 |
1mg |
£12060.00
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073324
|
Invitrogen™
PV5126 |
20 μg |
£564.00
Each |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PIK3C3 phosphorylates phosphatidylinositol (PI) to produce phosphatidylinositol 3-phosphate [PI(3)P], which recruits downstream effectors containing FYVE and Phox domains to membranes. PIK3C3 is involved in coupling nutrient signaling to mTOR and also plays a role in autophagy and intracellular trafficking.
Specifications
NP_002638.2 | |
PI3 Kinase Family | |
Recombinant | |
Baculovirus |
PIK3C3 (hVPS34) | |
Human | |
GST Tag |