missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Triethylamine (TEA) Solvent

Bottles of 99.5%-pure TEA (MW 101.2, CAS 121-44-8) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.

Brand:  Thermo Scientific™ TS-25108

Code : NEW

Additional Details : Weight : 0.30000kg Transport : UN number : 1296 Chem class : 1b Pack group : II

 View more versions of this product

Product Code. 10471875

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Description

Description

Triethylamine is an ion-pairing reagent that alters selectivity in reverse-phase HPLC separations. By pairing with peptides, it effectively sharpens peaks, resulting in improved peak resolution.

Highlights:

  • >99.5% triethylamine purity, allowing sensitive peptide detection at low UV wavelengths in reverse-phase HPLC peptide separation systems Packaged in amber glass bottles with protective PTFE-lined fluorocarbon caps for reagent integrity
  • Has a low UV absorbance to provide the most sensitive detection across all wavelengths

Properties of Triethylamine

  • Alternate names TEA, Diethylethanamine Molecular formula C6H15N Molecular weight 101.19 Density 0.726g/mL

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.

Specifications

Specifications

HPLC
Flammable, Harmful, Irritant
Triethylamine
Pierce Triethylamine (TEA), Sequencing Grade
100g
2° - 8°C
100g
98 to 100%
Clear, colorless liquid of TEA (at least 99.5% pure; low water)
Videos