Learn More
Thermo Scientific™ Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format
Prepare amine-reactive sulfo-NHS-ester crosslinkers or enhance carbodiimide (EDC) conjugation for peptides or proteins using N-hydroxysulfosuccinimide, CAS 106627-54-7.
Brand: Thermo Scientific™ 24525
1402.20 GBP valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Code : PN
Additional Details : CAS Number : 106627-54-7 Weight : 0.45360kg
Description
Thermo Scientific™ Pierce Sulfo-NHS is pure N-hydroxysulfosuccinimide, the essential compound needed to prepare water-soluble, amine-reactive NHS-ester crosslinkers or to enhance conjugation efficiencies when using the carbodiimide EDC.
Highlights:
- Forms an acyl amino ester with carboxyl groups when used with the EDC
- Sulfo-NHS-activated molecule reacts efficiently with primary amines, resulting in stable amide bond
- Allows for two-step EDC reactions between carboxyl- and amine-containing molecules
- Sulfo-NHS-activated molecule can be stabilized by drying, allowing manufacture of amine-reactive probes and surfaces
- Reactive groups: Sulfo-NHS
- Reactive toward: amino groups (upon activation of carboxylates)
Specifications
Amine-Carboxyl | |
Sulfo-NHS | |
No | |
No | |
Pierce™ | |
Heterobifunctional | |
Short (<10 Å) | |
5 g | |
Standard, Single-use, Premium-grade | |
Crosslinker |
No | |
217.13 | |
Upon receipt store at 4°C. | |
Ambient | |
Chemical Labeling | |
Carbodiimide | |
Powder | |
Water | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.