Learn More
Thermo Scientific™ Pierce™ Sulfo-NHS Crosslinker
Prepare amine-reactive sulfo-NHS-ester crosslinkers or enhance carbodiimide (EDC) conjugation for peptides or proteins using N-hydroxysulfosuccinimide, CAS 106627-54-7.
Brand: Thermo Scientific™ 24520
Code : PN
Additional Details : Weight : 0.80000kg
Description
Highlights:
- Forms an acyl amino ester with carboxyl groups when used with the EDC
- Sulfo-NHS-activated molecule reacts efficiently with primary amines, resulting in stable amide bond
- Allows for two-step EDC reactions between carboxyl- and amine-containing molecules
- Sulfo-NHS-activated molecule can be stabilized by drying, allowing manufacture of amine-reactive probes and surfaces
- Reactive groups: Sulfo-NHS
- Reactive toward: amino groups (upon activation of carboxylates)
Specifications
Carboxyl-to-Amine | |
N-Hydroxysulfosuccinimide; solid | |
217.13 | |
8 x 2mg | |
0Å | |
Yes |
Pierce Sulfo-NHS, No-Weigh Format | |
Yes | |
No | |
15mL of typical 1X (5mM) reaction solution (total), or 8 separate uses | |
Store as detailed on product information sheet. |