Learn More
Thermo Scientific™ Sulfo-MBS (m-maleimidobenzoyl-N-hydroxysulfosuccinimide ester)
Crosslink amines to sulfhydryls with this short, water-soluble crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of an aromatic, 6-atom (7.3A) spacer arm.
Brand: Thermo Scientific™ 22312
Code : PN
Additional Details : CAS Number : 79-06-1 Weight : 0.80000kg
Description
Thermo Scientific™ Pierce Sulfo-MBS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short aromatic spacer arm (7.3 Ås).
Highlights:
- Water-soluble; membrane-impermeable
- Reactive groups: sulfo-NHS ester and maleimide
- Reactive toward: amino and sulfhydryl groups
Specifications
Amine-Sulfhydryl | |
Sulfo-MBS | |
No | |
Upon receipt store desiccated at -20°C. | |
Ambient | |
Chemical Labeling | |
Maleimide, Sulfo-NHS Ester | |
Powder | |
Water | |
Yes |
No | |
416.29 | |
7.3 Å | |
No | |
Pierce™ | |
Heterobifunctional | |
Short (<10 Å) | |
50 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.