Learn More
Thermo Scientific™ Pierce™ SILAC Media and Sera
Analyze protein expression by mass spectrometry using these media or additives for stable isotope labeling with amino acids in cell culture (SILAC).
£39.03 - £443.88
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
10404205
|
Thermo Scientific™
88215 |
DMEM:F12 (1:1) media minus L-Lysine and L-Arginine for induced pluripotent cells | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12116490
|
Thermo Scientific™
88440 |
Dialyzed fetal bovine serum | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10423425
|
Thermo Scientific™
89984 |
RPMI-1640 media minus L-Lysine and L-Arginine | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10107883
|
Thermo Scientific™
89985 |
DMEM media minus L-Lysine and L-Arginine | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10511404
|
Thermo Scientific™
89986 |
Dialyzed fetal bovine serum | 50 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
These Thermo Scientific Pierce cell culture media and serum components are optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).
SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of heavy 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.Highlights:
Flexible–liquid media deficient in both L-lysine and L-arginine and powdered media deficient in L-leucine, L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
Compatible–DMEM, MEM, RPMI-1640, IMDM, Ham's F12 and McCoy's 5A media and dialyzed FBS are used to culture a variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3, K562 and Jurkat
Optimized, specialized media–the Expansion DMEM and Low-Osmolarity DMEM are specifically designed and qualified for mouse embryonic stem cell propagation and SILAC protein expression profiling with mass spectrometry
High quality–Stem Cell Screened Dialyzed FBS is tested to ensure that it does not promote stem cell differentiation