Learn More
Thermo Scientific™ Pierce™ RED Device for Rapid Equilibrium Dialysis
Perform plasma protein-binding assays, a critical step in drug development, with the easiest, fastest and most reliable rapid equilibrium dialysis (RED) system.
£33.66 - £3125.00
Products 10
Product Code Brand Description Quantity MWCO Price Quantity & Availability  
Product Code Brand Description Quantity MWCO Price Quantity & Availability  
10250544 View Documents Thermo Scientific™
89809
Pierce RED Device Inserts 50 inserts 8000Da £364.00
50 units View alternative products
Save Up to 
»
EU Stock 26 Please to purchase this item. Need a web account? Register with us today!
11864131 View Documents Thermo Scientific™
89810
Pierce RED Device Inserts 250 inserts 8000Da £1508.00
250 units View alternative products
Save Up to 
»
EU Stock 24 Please to purchase this item. Need a web account? Register with us today!
10731585 View Documents Thermo Scientific™
89811
Pierce RED Device Reusable Base Plate 1 plate ---- £343.00
Each View alternative products
Save Up to 
»
EU Stock 5 Please to purchase this item. Need a web account? Register with us today!
10568913 View Documents Thermo Scientific™
89812
Pierce RED Device Insert Removal Tool 1 tool ---- £86.00
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-03-2020
11884191 View Documents Thermo Scientific™
90004
Pierce RED Device Single-Use Base Plates 2 plates ---- £33.66
Pack of 2 View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
10066903 View Documents Thermo Scientific™
90006
Pierce RED Device Single-Use Plate with Inserts 1 plate set 8000Da £422.00
Each View alternative products
Save Up to 
»
EU Stock 11 Please to purchase this item. Need a web account? Register with us today!
11804201 View Documents Thermo Scientific™
90007
Pierce RED Device Single-Use Plate with Inserts 5 plate set 8000Da £1688.00
Pack of 5 View alternative products
Save Up to 
»
In Stock 2
EU Stock 2
12311963 View Documents Thermo Scientific™
90112
Pierce RED Device Single-Use Plate with Inserts 1 plate set 12000Da £424.00
1 set View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
12201303 View Documents Thermo Scientific™
91012
Pierce RED Device Single-Use Plate with Inserts 10 plate set 12000Da £3125.00
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
11894191 View Documents Thermo Scientific™
90005
Pierce RED Device Single-Use Base Plates 10 plates ---- £148.00
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Description
Description
The RED Device is an apparatus for performing equilibrium dialysis experiments in a high throughput, automation-compatible format. The device consists of disposable inserts and a base plate formatted to a standard microplate footprint.
Each single-use, disposable insert is made of two side-by-side chambers separated by a vertical cylinder of dialysis membrane (8K MWCO) validated for minimal nonspecific binding. This format requires no extensive assembly steps or specialized equipment, and each chamber or well is easily accessible from the top of the insert after insertion in the base plate. Additionally, the high surface-to-volume ratio of the membrane compartment allows rapid dialysis, where equilibrium can be reached in 4 hours with high levels of reproducibility and accuracy. In many cases, experiments can be completed in less than 100 minutes.
RED Device Inserts are designed to be used with either the reusable PTFE or disposable high-density polypropylene base plates. Each RED Device Base Plate holds up to 48 RED Device Inserts and has a standard 96-well plate footprint with 9mm x 9mm well spacing to provide compatibility with automated liquid handling systems. In addition, the disposable RED Device Base Plates are available pre-loaded, providing operation convenience for scientists conducting protein-binding applications. No pre-conditioning of the membrane inserts is needed. When using radioactive materials, this single-use plate is easily disposed of to avoid contamination and cleaning. RED Device Inserts and Base Plates are also available separately.
Highlights:
  • Easy and ready to use – disposable tubes require no presoaking, assembly or specialized equipment
  • Designed for speed – the high surface-to-volume ratio of the insert design enables equilibrium to be reached in as few as 100 minutes with vigorous agitation or in three to four hours with 200rpm agitation
  • Automation-compatible – designed on a standard 96-well plate template suitable for automated liquid handlers
  • Flexible and scalable – perform any number of assays (1 to 48 assays per plate) without wasting the entire plate
  • Robust – compartmentalized design eliminates potential for cross talk or leakage
  • Reproducible and accurate – validated for plasma binding assays, producing results consistent with those reported in literature
  • Quality-tested – each lot of inserts is functionally tested in a protein-binding assay for guaranteed performance
Insert Features:
  • Disposable: require no presoaking, assembly, or specialized equipment
  • Short incubation time: large dialysis surface area accelerates equilibrium
  • 8K MWCO membrane: ideal molecular-weight cutoff for protein-drug binding studies
  • Membrane composition: regenerated cellulose with low glycerol content as a humectant
Base Plate Features:
  • Microplate footprint: compatible with automated systems for 96-well plates
  • Compartmentalized: eliminates potential for crosstalk or leakage
  • PTFE construction: eliminates nonspecific binding and risk of contamination
  • Accepts 1 to 48 inserts: run exactly the number of assays needed without waste
Recommended for:
Plasma protein-binding assays; Drug partition between plasma and whole blood; Determination of protein binding of liver microsomes to improve the correlation between in vitro and in vivo intrinsic clearance; Drug binding competition between tissues vs. plasma proteins; Pharmacokinetics studies; Formulation of drug dosage for in vivo studies; Drug-drug interaction studies; Selection criteria during drug lead optimization; Solubility studies; Dissociation constant determination (Kd); Tissue binding studies using tissue homogenate
Patent pending.