Thermo Scientific Pierce™ qIP Protein Interaction Assay Reagents

Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with these reagents.

Overview
Brand: Thermo Scientific

Manufacturer Part Number: 82014

X10 QIP PROTEIN INTERACTION BUFFER D

UNSPSC: 41116010

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 15255693

Quantity Price
1 £ 110.0 / 50mL
Estimated Shipment date
from Supplier 10-03-2017
Add to basket


Description and Specification

Specification

Quantity 50mL
Formulation Proprietary aqueous buffer
Quantity 50mL
Sufficient For 500mL 1X Buffer D to dilute lysates and prepare wash buffer for qIP assays