Learn More
Thermo Scientific™ Pierce™ qIP Protein Interaction Assay Reagents
Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with these reagents.
Brand:  Thermo Scientific™ 82016
Code : NEW
Additional Details : Weight : 0.25000kg Transport : UN number : 1992 Chem class : 1 Pack group : II
 View more versions of this product
Product Code. 15215703
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Two-component kit
Coelenterazine Substrate (100X), 2 x 0.5mL
1500-rxn kit
1500 luciferase assay reactions
Videos